Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Centrifuge 5702 G
Synonym(s):
Eppendorf
®
Centrifuge 5702 R G
Products
Product Category
centrifuges (4)
Manufacturer
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP5702000010
Eppendorf
®
Centrifuge 5702 G,
w/o rotor, AC/DC input 230 V, 50 - 60 Hz
Expand
View Pricing
EP5702000065
Eppendorf
®
Centrifuge 5702 G,
w/o rotor, AC/DC input 230 V, 50 - 60 Hz, AC/DC input, UK plug
Expand
View Pricing
EP5702000320
Eppendorf
®
Centrifuge 5702 G,
incl. rotor A-4-38 and 15/50 mL adapters, AC/DC input 230 V, 50 - 60 Hz
Expand
View Pricing
EP5702000365
Eppendorf
®
Centrifuge 5702 G,
AC/DC input 230 V, 50 - 60 Hz, incl. rotor A-4-38 and 15/50ml adapters, AC/DC input, UK plug
Expand
View Pricing