Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Empore
™
SPE Disks
Products
Product Category
disks (10)
Brand
Supelco (7)
Manufacturer
Product Line
Chemical Composition
Diameter (mm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
66883-U
Empore
™
SPE Disks,
C18, diam. 47 mm, pk of 20
Expand
View Pricing
66884-U
Empore
™
SPE Disks,
polystyrene-divinylbenzene (SDB-XC), diam. 47 mm, pk of 20
Expand
View Pricing
66896-U
Empore
™
SPE Disks,
Carbon, diam. 47 mm, pk of 20
Expand
View Pricing
66886-U
Empore
™
SPE Disks,
SDB-RPS, diam. 47 mm, pk of 20
Expand
View Pricing
66882-U
Empore
™
SPE Disks,
C8, diam. 47 mm, pk of 20
Expand
View Pricing
66889-U
Empore
™
SPE Disks,
Cation Exchange, diam. 47 mm, pk of 20
Expand
View Pricing
66894-U
Empore
™
SPE Disks,
Chelating resin, diam. 47 mm, pk of 20
Expand
View Pricing
66888-U
Empore
™
SPE Disks,
Anion Exchange, pk of 20, diam. 47 mm
Expand
View Pricing
66887-U
Empore
™
SPE Disks,
Oil and Grease, diam. 47 mm, pk of 20
Expand
View Pricing
66898-U
Empore
™
SPE Disks,
Oil and Grease, diam. 90 mm, pk of 10
Expand
View Pricing