
Dyglycine

Synonym(s):
N-Glycylglycine
Empirical Formula (Hill Notation):
C4H8N2O3
CAS Number:
Molecular Weight:
132.12
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing