Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(+)-Valencene
Synonym(s):
(3
R
,4
aS
,5
R
)-4
a
,5-Dimethyl-3-isopropenyl-1,2,3,4,4
a
,5,6,7-octahydronaphthalene
Empirical Formula (Hill Notation):
C
15
H
24
CAS Number:
4630-07-3
Molecular Weight:
204.35
EC Number:
225-047-6
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technical (1)
analytical standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
75056
(+)-Valencene,
technical, ≥70%
Expand
View Pricing
06808
(+)-Valencene,
analytical standard
Expand
View Pricing