Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Laboratory Bottle Wide-Neck Amber
Products
Product Category
bottles (4)
Capacity (mL)
Cap
Color
Vial Bottle Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
DWK218663637
Duran
®
Laboratory Bottle Wide-Neck Amber,
amber bottle, protect, without cap, GLS 80 (without ring), capacity 250 mL
Expand
View Pricing
DWK218664436
Duran
®
Laboratory Bottle Wide-Neck Amber,
amber bottle, protect, without cap, GLS 80 (without ring), capacity 500 mL
Expand
View Pricing
DWK218666334
Duran
®
Laboratory Bottle Wide-Neck Amber,
amber bottle, protect, without cap, GLS 80 (without ring), capacity 2000 mL
Expand
View Pricing
DWK218667339
Duran
®
Laboratory Bottle Wide-Neck Amber,
amber bottle, protect, without cap, GLS 80 (without ring), capacity 5000 mL
Expand
View Pricing