
Dulcoside A

Synonym(s):
(4α)-13-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester
Empirical Formula (Hill Notation):
C38H60O17
CAS Number:
Molecular Weight:
788.87
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing