Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(+)-Sodium L-ascorbate
Synonym(s):
L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt
Empirical Formula (Hill Notation):
C
6
H
7
NaO
6
CAS Number:
134-03-2
Molecular Weight:
198.11
EC Number:
205-126-1
Products
Product Category
vitamins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A7631
(+)-Sodium L-ascorbate,
crystalline, ≥98%
Expand
View Pricing
11140
(+)-Sodium L-ascorbate,
BioXtra, ≥99.0% (NT)
Expand
View Pricing
A4034
(+)-Sodium L-ascorbate,
powder, BioReagent, suitable for cell culture
Expand
View Pricing