
DMT-dG(dmf) Phosphoramidite

Synonym(s):
DMT-dG(dmf) Amidite, N-[(dimethylamino)methylene]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N2-Dimethylformamidine-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]
Empirical Formula (Hill Notation):
C43H53N8O7P
CAS Number:
Molecular Weight:
824.90
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing