
(±)-Methadone-D9 solution

Empirical Formula (Hill Notation):
C21D9H18NO
Molecular Weight:
318.50
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique