Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Discharge valve for Dispensette
®
S
Products
Product Category
valves (7)
Manufacturer
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BR6727
Discharge valve for Dispensette
®
S,
5 and 10 mL, (PFA/Boro 3.3/ceramic/platinum-iridium), no valve marking
Expand
View Pricing
BR6728
Discharge valve for Dispensette
®
S,
25, 50 and 100 mL, (PFA/Boro 3.3/ceramic/platinum-iridium), no valve marking
Expand
View Pricing
BR6729
Discharge valve for Dispensette
®
S,
Organic, 5 and 10 mL, (PFA/Boro 3.3/ceramic/tantalum), valve marking with "ORG"
Expand
View Pricing
BR6730
Discharge valve for Dispensette
®
S,
Organic, 25, 50 and 100 mL, (PFA/Boro 3.3/ceramic/tantalum), valve marking with "ORG"
Expand
View Pricing
BR6732
Discharge valve for Dispensette
®
S,
Trace Analysis, 10 mL, PT/IR spring, valve marking with ′Pt-Ir′
Expand
View Pricing
BR6749
Discharge valve for Dispensette
®
S,
1 and 2 mL, (PFA/Boro 3.3/ceramic/platinum-iridium), no valve marking
Expand
View Pricing
BR6733
Discharge valve for Dispensette
®
S,
Trace Analysis, 10 mL, Tantalum spring, valve marking with ′Ta′
Expand
View Pricing