Skip to Content
Merck
Diphenyl ether for ion-selective electrodes, Selectophore™, ≥99.9%

Diphenyl ether

Diphenyl ether for ion-selective electrodes, Selectophore™, ≥99.9%
Synonym(s):
Diphenyl oxide, Phenyl ether, Diphenyl ether
Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade