Dihydrocodeine-D6 hydrochloride solution
Empirical Formula (Hill Notation):
C18H17D6NO3·HCl
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing