Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethyl carbonate
Synonym(s):
Diethyl carbonate
Linear Formula:
CO(OC
2
H
5
)
2
CAS Number:
105-58-8
Molecular Weight:
118.13
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (4)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
D91551
Diethyl carbonate,
99%
Expand
View Pricing
517135
Diethyl carbonate,
anhydrous, ≥99%
Expand
View Pricing
900018
Diethyl carbonate,
battery grade, ≥99%, acid <10 ppm, H2O <10 ppm
Expand
View Pricing
8.02898
Diethyl carbonate,
for synthesis
Expand
View Pricing