
Demeclocycline hydrochloride

Synonym(s):
7-Chloro-6-demethyltetracycline hydrochloride, Demethylchlorotetracycline hydrochloride, Demeclocycline hydrochloride
Empirical Formula (Hill Notation):
C21H21ClN2O8 · HCl
CAS Number:
Molecular Weight:
501.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Research Area
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing