
D-α-(4-Hydroxy-phenyl)glycine

Synonym(s):
4-Hydroxy-D-phenylglycine, (2R)-2-Amino-2-(4-hydroxyphenyl)acetic acid
Linear Formula:
HOC6H4CH(NH2)CO2H
CAS Number:
Molecular Weight:
167.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing