Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Thermowell GOLD PCR tubes
Products
Product Category
tubes (6)
Color
Manufacturer
Capacity (mL)
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3745
Corning
®
Thermowell GOLD PCR tubes,
0.2mL PCR tubes, flat cap, non-sterile, clear, 1000/cs
Expand
View Pricing
CLS3744
Corning
®
Thermowell GOLD PCR tubes,
0.2mL PCR tubes, flat cap, non-sterile, assorted colors, 1000/cs
Expand
View Pricing
CLS3749
Corning
®
Thermowell GOLD PCR tubes,
0.5mL PCR tubes, flat cap, non-sterile, assorted colors, 1000/cs
Expand
View Pricing
CLS3750
Corning
®
Thermowell GOLD PCR tubes,
0.5mL PCR tubes, flat cap, non-sterile, clear, 1000/cs
Expand
View Pricing
CLS3747
Corning
®
Thermowell GOLD PCR tubes,
0.2mL PCR tubes, domed cap, non-sterile, clear, 1000/cs
Expand
View Pricing
CLS3746
Corning
®
Thermowell GOLD PCR tubes,
0.2mL PCR tubes, domed cap, non-sterile, assorted colors, 1000/cs
Expand
View Pricing