Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
centrifuge bottle
Products
Product Category
bottles (7)
Manufacturer
Capacity (mL)
Cap
Color
Vial Bottle Color
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS431845
Corning
®
centrifuge bottle,
with standard screw cap, capacity 500 mL, polypropylene
Expand
View Pricing
CLS431840
Corning
®
centrifuge bottle,
with silicon O-ring screw cap, capacity 250 mL, polypropylene
Expand
View Pricing
CLS431842
Corning
®
centrifuge bottle,
with silicon O-ring screw cap, capacity 250 mL, polycarbonate
Expand
View Pricing
CLS431846
Corning
®
centrifuge bottle,
with silicon O-ring screw cap, capacity 500 mL, polycarbonate
Expand
View Pricing
CLS431844
Corning
®
centrifuge bottle,
with silicon O-ring screw cap, capacity 500 mL, polypropylene
Expand
View Pricing
CLS431847
Corning
®
centrifuge bottle,
with standard screw cap, capacity 500 mL, polycarbonate
Expand
View Pricing
CLS431843
Corning
®
centrifuge bottle,
with standard screw cap, capacity 250 mL, polycarbonate
Expand
View Pricing