Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
BioCoat
™
Collagen I Microplate
Products
Product Category
plates (22)
Manufacturer
Binding Type
Wells
Color
Sterility
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS354407
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356667
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear black wells, lid, sterile
Expand
View Pricing
CLS356664
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear white wells, lid, sterile
Expand
View Pricing
CLS356519
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom opaque white wells, lid
Expand
View Pricing
CLS356649
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear black wells, lid
Expand
View Pricing
CLS356700
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear black wells
Expand
View Pricing
CLS354649
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear black wells, lid
Expand
View Pricing
CLS354519
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom opaque white wells, lid
Expand
View Pricing
CLS356699
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom white wells
Expand
View Pricing
CLS354664
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear white wells, lid, sterile
Expand
View Pricing
CLS354666
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear wells, lid, sterile
Expand
View Pricing
CLS354667
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear black wells, lid
Expand
View Pricing
CLS356650
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear white wells, lid
Expand
View Pricing
CLS356665
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom white wells, lid, sterile
Expand
View Pricing
CLS356407
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear wells, lid
Expand
View Pricing
CLS356701
Corning
®
BioCoat
™
Collagen I Microplate,
well size 96 wells, flat bottom clear white wells
Expand
View Pricing
CLS354665
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom white wells, lid, sterile
Expand
View Pricing
CLS356702
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear wells
Expand
View Pricing
CLS356703
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom white wells
Expand
View Pricing
CLS356705
Corning
®
BioCoat
™
Collagen I Microplate,
well size 384 wells, flat bottom clear black wells
Expand
View Pricing
Page 1 of 2
Page 1 of 2