Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF03745795
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
100mm rod, 6mm diameter
Expand
View Pricing
GF10522011
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
100mm rod, 25mm diameter
Expand
View Pricing
GF41656237
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
200mm rod, 6mm diameter
Expand
View Pricing
GF37027627
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
500mm rod, 25mm diameter
Expand
View Pricing
GF07557453
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
500mm rod, 6mm diameter
Expand
View Pricing
GF50979774
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
200mm rod, 25mm diameter
Expand
View Pricing
GF22278551
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
1000mm rod, 6mm diameter
Expand
View Pricing
GF41235443
Copper-cobalt-beryllium alloy, Cu97Co2.5Be0.5,
1000mm rod, 25mm diameter
Expand
View Pricing