Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt/Silicon/Boron/Iron/Nickel
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF11939014
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.025 mm, length 2 m, coil width 25 mm
Expand
View Pricing
GF48868172
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.018 mm, length 1 m, coil width 50.8 mm
Expand
View Pricing
GF69695511
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co66%/Si15%/B 14%/Fe 4%/Ni 1%, thickness 0.025 mm, length 1 m, coil width 25 mm
Expand
View Pricing
GF92376445
Cobalt/Silicon/Boron/Iron/Nickel,
foil, thickness 0.025 mm, Co66%/Si15%/B 14%/Fe 4%/Ni 1%, length 5 m, coil width 25 mm
Expand
View Pricing
GF07846511
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.018 mm, length 10 m, coil width 50.8 mm
Expand
View Pricing
GF68193490
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co66%/Si15%/B 14%/Fe 4%/Ni 1%, thickness 0.018 mm, length 0.5 m, coil width 50.8 mm
Expand
View Pricing
GF99757568
Cobalt/Silicon/Boron/Iron/Nickel,
foil, thickness 0.025 mm, Co66%/Si15%/B 14%/Fe 4%/Ni 1%, length 0.5 m, coil width 25 mm
Expand
View Pricing
GF08886662
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.018 mm, length 2 m, coil width 50.8 mm
Expand
View Pricing
GF47055955
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.025 mm, length 10 m, coil width 25 mm
Expand
View Pricing
GF09356014
Cobalt/Silicon/Boron/Iron/Nickel,
foil, Co 66%/Si 15%/B 14%/Fe 4%/Ni 1%, thickness 0.018 mm, length 5 m, coil width 50.8 mm
Expand
View Pricing