Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Clear-view
™
Snap-Cap microtubes
Products
Product Category
tubes (4)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Capacity (mL)
Color
Package Size
Special Grade
Natural (3)
Sterility
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
T4816
Clear-view
™
Snap-Cap microtubes,
capacity 1.5 mL, natural, low retention
Expand
View Pricing
T3566
Clear-view
™
Snap-Cap microtubes,
size 1.5 mL, natural
Expand
View Pricing
T2441
Clear-view
™
Snap-Cap microtubes,
capacity 0.6 mL, natural
Expand
View Pricing
T4691
Clear-view
™
Snap-Cap microtubes,
capacity 0.6 mL
Expand
View Pricing