
Citalopram Related Compound C

Synonym(s):
3-(3-DIMETHYLAMINOPROPYL)-3-(4-FLUOROPHENYL)-6-CYANO-1(3H)-ISOBENZOFURANONE OXALATE
Empirical Formula (Hill Notation):
C22H21FN2O6
Molecular Weight:
428.41
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2