Skip to Content
Merck
cis-7,10,13,16-Docosatetraenoic acid ≥98% (GC)

cis-7,10,13,16-Docosatetraenoic acid

cis-7,10,13,16-Docosatetraenoic acid ≥98% (GC)
Synonym(s):
DTA
Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing