Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloroform solution
Products
Product Category
stable isotopes (5)
solvents (3)
pesticides (1)
Brand
Sigma-Aldrich (5)
Supelco (6)
Physical Form
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (4)
certified reference material (1)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
40021
Chloroform solution,
certified reference material, 5000 μg/mL in methanol
Expand
View Pricing
708976
Chloroform solution,
suitable for NMR (reference standard), 3% in acetone-d
6
(99.9 atom % D)
Expand
View Pricing
733814
Chloroform solution,
suitable for NMR (reference standard), 10% in acetone-d
6
(99.9 atom % D), NMR tube size 3 mm × 8 in.
Expand
View Pricing
612987
Chloroform solution,
suitable for NMR (reference standard), 3% in acetone-d
6
(99.9 atom % D), TMS 0.2 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
612979
Chloroform solution,
suitable for NMR (reference standard), 1% in acetone-d
6
(99,9 atom % D), TMS 0.1 %, NMR tube size 5 mm × 8 in.
Expand
View Pricing
720992
Chloroform solution,
suitable for NMR (reference standard), 0.3% in acetone-d
6
(99.9 atom % D), NMR tube size 3 mm × 8 in.
Expand
View Pricing
717916
Chloroform solution,
suitable for NMR (reference standard), 5% in acetone-d
6
(99.9 atom % D), chromium(III) acetylacetonate 0.2 %
Expand
View Pricing
717908
Chloroform solution,
suitable for NMR (reference standard), 50% in acetone-d
6
(99.9 atom % D), chromium(III) acetylacetonate 0.2 %
Expand
View Pricing
487163
Chloroform solution,
suitable for NMR (reference standard), 1% in acetone-d
6
(99.9 atom % D)
Expand
View Pricing
611859
Chloroform solution,
suitable for NMR (reference standard), 20% in acetone-d
6
(99.9 atom % D), NMR tube size 5 mm × 8 in.
Expand
View Pricing
611778
Chloroform solution,
suitable for NMR (reference standard), 1% in acetone-d
6
(99.9 atom % D), NMR tube size 3 mm × 8 in.
Expand
View Pricing