
Cesium methanesulfonate

Synonym(s):
CsMSF, CsMeS, Methanesulfonic acid cesium salt
Linear Formula:
CH3O3SCs
CAS Number:
Molecular Weight:
228.00
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing