
Cerium(IV) oxide

Synonym(s):
Ceric oxide, Ceric oxide, ceria, Cerium(IV) oxide
Linear Formula:
CeO2
CAS Number:
Molecular Weight:
172.11
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core
Special Grade
Showing 1-6 of 6
Compare
Product Number
Description
Pricing