Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cephalexin hydrate
Empirical Formula (Hill Notation):
C
16
H
17
N
3
O
4
S · xH
2
O
CAS Number:
1820673-23-1
Molecular Weight:
347.39 (anhydrous basis)
EC Number:
239-773-6
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
pK Value
Activity Spectrum
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C4895
Cephalexin hydrate,
first-generation cephalosporin antibiotic
Expand
View Pricing