Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Centrifuge Tube
Products
Product Category
tubes (7)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Special Grade
Natural (4)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
HS4427R
Centrifuge Tube,
capacity 50 mL, natural, conical bottom, pack of 500 ea (Racked), sterile
Expand
View Pricing
HS4429
Centrifuge Tube,
tube capacity 50 mL, black, conical bottom, pack of 500 ea (Bulk Wrap), sterile
Expand
View Pricing
HS4427B
Centrifuge Tube,
capacity 50 mL, natural, conical bottom, pack of 500 ea (Bulk Wrap)
Expand
View Pricing
HS4426B
Centrifuge Tube,
capacity 15 mL, conical bottom, pack of 500 ea (Bulk Wrap)
Expand
View Pricing
HS4428
Centrifuge Tube,
capacity 15 mL, black, conical bottom, pack of 500 ea (Bulk Wrap)
Expand
View Pricing
HS4426R
Centrifuge Tube,
capacity 15 mL, natural, conical bottom, pack of 500 ea (Racked)
Expand
View Pricing
HS4427S
Centrifuge Tube,
capacity 50 mL, natural, skirted bottom, pack of 500 ea (Bulk Wrap), sterile
Expand
View Pricing