Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon-
13
C monoxide-
18
O Gas
Linear Formula:
13
C
18
O
CAS Number:
35907-63-2
Molecular Weight:
31.00
Products
Product Category
building blocks (2)
gases (2)
stable isotopes (2)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
607142
Carbon-
13
C monoxide-
18
O Gas,
95 atom %
18
O, 99 atom %
13
C
Expand
View Pricing
655007
Carbon-
13
C monoxide-
18
O Gas,
99 atom %
18
O, 99 atom %
13
C
Expand
View Pricing