Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Camphor (dl)
Synonym(s):
(±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
Empirical Formula (Hill Notation):
C
10
H
16
O
CAS Number:
76-22-2
Molecular Weight:
152.23
EC Number:
200-945-0
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
primary reference standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
02070595
Camphor (dl),
primary reference standard
Expand
View Pricing