Skip to Content
Merck
Brij® 58 for synthesis

Brij® 58

Brij® 58 for synthesis
Synonym(s):
Polyethylene glycol hexadecyl ether, Polyoxyethylene (20) cetyl ether, Brij® 58
Linear Formula:
HO-(CH2CH2O)n-(CH2)15-CH3
CAS Number:

Products

Product Category
Brand
Melting Point (°C)
Physical Form
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing