Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX)
Products
Product Category
pipettes (33)
Manufacturer
Agency
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BR27819
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 2 mL, type 2
Expand
View Pricing
Z325600
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 0.5 mL , with 0.01 subdivisions
Expand
View Pricing
Z325724
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 10 mL , with 0.1 subdivisions, suitable for cotton plugs
Expand
View Pricing
BR27823
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 10 mL , with 0.1 mL subdivisions, type 2, suitable for cotton plugs
Expand
View Pricing
BR27816
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 0.5 mL , with 0.01 mL subdivisions, type 2
Expand
View Pricing
Z325848
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 5 mL , with 0.05 subdivisions, suitable for cotton plugs
Expand
View Pricing
Z325643
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 1 mL , with 0.01 subdivisions, suitable for cotton plugs
Expand
View Pricing
BR27817
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 1 mL , with 0.01 mL subdivisions, type 2
Expand
View Pricing
Z325805
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 1 (zero on top), capacity 1 mL , with 0.01 subdivisions
Expand
View Pricing
BR27826
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 50 mL , with 0.5 mL subdivisions, type 2, suitable for cotton plugs
Expand
View Pricing
Z325856
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 1 (zero on top), camber 10 mL , with 0.1 subdivisions, suitable for cotton plugs
Expand
View Pricing
Z325732
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
camber 20 mL, subdivision, 0.1 mL, suitable for cotton plugs
Expand
View Pricing
BR27825
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
capacity 25 mL , with 0.1 mL subdivisions, type 2, suitable for cotton plugs
Expand
View Pricing
BR27818
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
camber 1 mL , with 0.1 mL subdivisions, type 2
Expand
View Pricing
Z325708
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 5 mL , with 0.05 subdivisions, suitable for cotton plugs
Expand
View Pricing
Z325791
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 1 (zero on top), capacity 0.5 mL , with 0.01 subdivisions
Expand
View Pricing
BR27821
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
volume 5 mL , with 0.05 mL subdivisions, type 2, suitable for cotton plugs
Expand
View Pricing
Z325740
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 25 mL , with 0.1 subdivisions, suitable for cotton plugs
Expand
View Pricing
Z325716
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 3 (zero on top), capacity 5 mL , with 0.1 subdivisions, suitable for cotton plugs
Expand
View Pricing
Z325864
BRAND
®
BLAUBRAND
®
graduated pipette, calibrated to deliver (TD, EX),
type 1 (zero on top), capacity 25 mL , with 0.1 subdivisions, suitable for cotton plugs
Expand
View Pricing
Page 1 of 2
Page 1 of 2