
Boscalid

Synonym(s):
2-Chloro-N-(4′-chlorobiphenyl-2- yl)-nicotinamide
Empirical Formula (Hill Notation):
C18H12Cl2N2O
CAS Number:
Molecular Weight:
343.21
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2