
BIS-TRIS hydrochloride

Synonym(s):
BIS-TRIS hydrochloride, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane
Empirical Formula (Hill Notation):
C8H19NO5 · HCl
CAS Number:
Molecular Weight:
245.70
Products
Product Category
Brand
Biological Source
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing