
Bisacodyl Related Compound C

Synonym(s):
4-[[4-(Acetyloxy)phenyl]-2-pyridinylmethyl]phenol, 4-[(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl acetate
Empirical Formula (Hill Notation):
C20H17NO3
CAS Number:
Molecular Weight:
319.35
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2