![Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer 96%](/deepweb/assets/sigmaaldrich/product/structures/700/585/b2e5ae1d-2b88-42c8-a071-ef828d4a104c/640/b2e5ae1d-2b88-42c8-a071-ef828d4a104c.png)
Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer
![Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer 96%](/deepweb/assets/sigmaaldrich/product/structures/700/585/b2e5ae1d-2b88-42c8-a071-ef828d4a104c/640/b2e5ae1d-2b88-42c8-a071-ef828d4a104c.png)
Synonym(s):
2,5-Norbornadiene-rhodium(I) chloride dimer, Chloro(2,5-norbornadiene)rhodium(I) dimer, Rhodium(I) chloride 2,5-Norbornadiene complex dimer, [Rh(nbd)Cl]2
Empirical Formula (Hill Notation):
C14H16Cl2Rh2
Products
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing