
Benzoyl chloride-d5

Synonym(s):
Pentadeuterobenzoyl chloride
Linear Formula:
C6D5COCl
CAS Number:
Molecular Weight:
145.60
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing