Skip to Content
Merck
Azomethine-H monosodium salt hydrate ~95%

Azomethine-H monosodium salt hydrate

Azomethine-H monosodium salt hydrate ~95%
Synonym(s):
4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2,7-disulfonic acid monosodium salt hydrate, 4-Hydroxy-5-(salicylideneamino)-2,7-naphthalenedisulfonic acid monosodium salt
Empirical Formula (Hill Notation):
C17H12NNaO8S2 · xH2O
CAS Number:
Molecular Weight:
445.40 (anhydrous basis)

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing