Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Axygen MAXYMum Recovery
®
tip
Products
Manufacturer
Color
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
AXYVT38410LR
Axygen MAXYMum Recovery
®
tip,
size 10 μL, clear
Expand
View Pricing
AXYVT38431LR
Axygen MAXYMum Recovery
®
tip,
fine point, size 30 μL
Expand
View Pricing
AXYVT38431LRS
Axygen MAXYMum Recovery
®
tip,
fine point, size 30 μL, sterile
Expand
View Pricing
AXYVTF38415LRS
Axygen MAXYMum Recovery
®
tip,
filtered, size 15 μL, clear, sterile
Expand
View Pricing
AXYVT38470LRS
Axygen MAXYMum Recovery
®
tip,
size 70 μL, clear, sterile
Expand
View Pricing
AXYVT3845LRS
Axygen MAXYMum Recovery
®
tip,
size 5 μL, clear, sterile
Expand
View Pricing
AXYVT38470LR
Axygen MAXYMum Recovery
®
tip,
size 70 μL, clear
Expand
View Pricing
AXYVT3845LR
Axygen MAXYMum Recovery
®
tip,
size 5 μL, clear
Expand
View Pricing