Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Avidin from egg white
CAS Number:
1405-69-2
EC Number:
215-783-6
Products
Product Category
proteins (1)
Brand
Sigma-Aldrich (2)
Physical Form
Product Line
Purity
Recombinant Host
Tissue
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A9275
Avidin from egg white,
BioUltra, lyophilized powder, ≥10 units/mg protein (E1%/280), ≥98% (SDS-PAGE)
Expand
View Pricing
A8706
Avidin from egg white,
recombinant, expressed in corn, ≥12 units/mg protein (E1%/280)
Expand
View Pricing