![Asperuloside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/102/009/caa7fc39-e799-486c-9a8d-3e84b05dd8cf/640/caa7fc39-e799-486c-9a8d-3e84b05dd8cf.png)
Asperuloside
![Asperuloside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/102/009/caa7fc39-e799-486c-9a8d-3e84b05dd8cf/640/caa7fc39-e799-486c-9a8d-3e84b05dd8cf.png)
Empirical Formula (Hill Notation):
C18H22O11
CAS Number:
Molecular Weight:
414.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing