![Anti-Human IgG4−FITC antibody, Mouse monoclonal clone HP-6025, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/128/404/8536659e-d6df-4dd6-881b-4c6e0c535230/640/8536659e-d6df-4dd6-881b-4c6e0c535230.jpg)
Anti-Human IgG4−FITC antibody, Mouse monoclonal
![Anti-Human IgG4−FITC antibody, Mouse monoclonal clone HP-6025, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/128/404/8536659e-d6df-4dd6-881b-4c6e0c535230/640/8536659e-d6df-4dd6-881b-4c6e0c535230.jpg)
Synonym(s):
Monoclonal Anti-Human IgG4
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing