
Anti-GLCCI1 antibody produced in rabbit

Synonym(s):
Anti-Glucocorticoid-induced transcript 1 protein antibody produced in rabbit, Anti-FAM117C, Anti-GIG18
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing