
Anti-Bovine IgG (whole molecule) antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing