Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Anise oil
Synonym(s):
Anise seed oil
CAS Number:
8007-70-3
Products
Product Category
essential oils (1)
esters (1)
lipids (1)
natural products (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
10521
Anise oil,
nature identical
Expand
View Pricing
1036959
Anise oil,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing