Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium iron(III) sulfate dodecahydrate
Synonym(s):
Ammonium ferric sulfate dodecahydrate, Ammonium iron(III) sulfate dodecahydrate
Linear Formula:
NH
4
Fe(SO
4
)
2
· 12H
2
O
CAS Number:
7783-83-7
Molecular Weight:
482.19
EC Number:
233-382-4
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
221260
Ammonium iron(III) sulfate dodecahydrate,
ACS reagent, 99%
Expand
View Pricing
1.03776
Ammonium iron(III) sulfate dodecahydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
F3629
Ammonium iron(III) sulfate dodecahydrate,
ReagentPlus
®
, ≥99%
Expand
View Pricing