
Amido Black Staining Solution 2X

Synonym(s):
Naphthol Blue Black solution, Amido Black 10B
Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing