Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Allopurinol
Synonym(s):
1
H
-Pyrazolo(3,4-d)pyrimidin-4-ol, 4-Hydroxypyrazolo(3,4-d)pyrimidine, 4-Hydroxypyrazolo[3,4-d]pyrimidine, HPP, Allopurinol
Empirical Formula (Hill Notation):
C
5
H
4
N
4
O
CAS Number:
315-30-0
Molecular Weight:
136.11
EC Number:
206-250-9
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Special Grade
pharmaceutical primary standard (3)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
A8003
Allopurinol,
xanthine oxidase inhibitor
Expand
View Pricing
1013002
Allopurinol,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1377
Allopurinol,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
A0350000
Allopurinol,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP870
Allopurinol,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing