Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Essentials Erlenmeyer flask, PTFE lined screw cap
Products
Product Category
flasks (4)
Brand
Sigma-Aldrich (4)
Capacity (mL)
Cap
Color
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z740668
Aldrich
®
Essentials Erlenmeyer flask, PTFE lined screw cap,
capacity 100 mL, 24-410 thread, volume 50-100 mL, accuracy: 25 mL
Expand
View Pricing
Z740665
Aldrich
®
Essentials Erlenmeyer flask, PTFE lined screw cap,
capacity 50 mL, 24-410 thread, volume 20-50 mL, accuracy: 10 mL
Expand
View Pricing
Z740677
Aldrich
®
Essentials Erlenmeyer flask, PTFE lined screw cap,
capacity 1000 mL, 38-430 thread, volume 250-1000 mL, accuracy: 50 mL
Expand
View Pricing
Z740674
Aldrich
®
Essentials Erlenmeyer flask, PTFE lined screw cap,
capacity 500 mL, 38-430 thread, volume 100-500 mL, accuracy: 50 mL
Expand
View Pricing