Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapt-a-Rack
®
Flexible Multi-Tube Rack
Products
Product Category
racks (8)
Color
Manufacturer
Diameter (mm)
Wells
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
HS120184
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, Combo Blue/Green, pk of 2
Expand
View Pricing
HS120187
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, blue, pk of 2
Expand
View Pricing
HS120188
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, white, pk of 2
Expand
View Pricing
HS120190
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, yellow, pk of 2
Expand
View Pricing
HS120185
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, Combo Blue/Yellow, pk of 2
Expand
View Pricing
HS120191
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, green, pk of 2
Expand
View Pricing
HS120186
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, Combo Pink/White, pk of 2
Expand
View Pricing
HS120189
Adapt-a-Rack
®
Flexible Multi-Tube Rack,
for tubes, 5 ‑ 50 mL, pink, pk of 2
Expand
View Pricing